Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, Ultrapure, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18607658 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
1 kg
5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-658 25 g Poly Bottle
18-607-664 125 g Poly Bottle
18-607-659 500 g Poly Bottle
18-607-660 1 kg Poly Bottle
18-607-661 5 kg Poly Pail
18-607-665 12 kg Poly Pail
18-607-662 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-658 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. U101625GM
Add to Cart
Add to Cart

U1016, 57-13-6, CH4N2O

Spectrum™ Chemical Urea, Ultrapure is an odorless and colorless solid that is an important nitrogen-containing substance found in mammal urine. Urea is an effective protein denaturant. In a highly concentrated (6 to 8 M) urea solutions, many proteins unfold and separate into their constituent polypeptide chains. Urea UltraPure is also used to denature nucleic acids in sequencing gels. Ultrapure reagents offered by Spectrum undergo rigorous specification testing to insure accurate, repeatable results.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O

Specifications

CAS Max % 1
Melting Point 132°C to 135°C
Packaging Poly Bottle
Quantity 25 g
Percent Purity 99.5%
Grade Ultrapure
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.