Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, USP, JP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607666 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-666 500 g Poly Bottle
18-607-667 2.5 kg Poly Pail
18-607-669 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-666 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. UR107500GM
Add to Cart
Add to Cart

UR107, 57-13-6, CH4N2O

Spectrum™ Chemical Urea, USP, JP is used in topical dermatological products to promote rehydration of the skin. Urea preparations are also used for non-surgical removal of damaged or dead nails. All Spectrum Chemical USP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98 to 102%
Grade Bio Certified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.