Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urease from Jack Bean, TCI America™
SDP

Catalog No. U00171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00171G 1 g
U0017100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00171G Supplier TCI America Supplier No. U00171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9002-13-5
MDL Number MFCD00070858

Specifications

Color White-Yellow
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Urease from Jack Bean
RTECSNumber YU1700000
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.