Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urethane, 97%

Catalog No. AC325540010
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325540010 1 kg Plastic bottle
AC325540500 50 g Plastic bottle
AC325542500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC325540010 Supplier Thermo Scientific Chemicals Supplier No. 325540010
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 51-79-6 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-79-6
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00007966
InChI Key JOYRKODLDBILNP-UHFFFAOYSA-N
Synonym urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
PubChem CID 5641
ChEBI CHEBI:17967
IUPAC Name ethyl carbamate
SMILES CCOC(=O)N

Specifications

Melting Point 47.0°C to 51.0°C
Color White
Boiling Point 182.0°C to 184.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula H2NCO2C2H5
Quantity 1 kg
Beilstein 03,22
Fieser 01,127
Merck Index 15,10059
Solubility Information Solubility in water: 200g/100 mL (20°C). Other solubilities: soluble in ethanol (125g/100 mL),chloroform,(111g/100 mL),ether (66.67g/100 mL),glycerol,(40g/100 mL),,slightly soluble in olive oil (3g/100 mL)
Formula Weight 89.09
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Urethane, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause cancer.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before us
EINECSNumber 200-123-1
RTECSNumber FA8400000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.