Learn More
Medchemexpress LLC HY-B2130 1g Medchemexpress, Uric acid CAS:69-93-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB21301G
Encompass_Preferred
Medchemexpress, HY-B2130 1g Uric acid CAS:69-93-2 Uric acid is an endogenous antioxidant that scavenges reactive oxygen species (ROS) including singlet oxygen, oxygen radicals, and peroxynitrite. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3074
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.