Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Uric Acid Dry Powder Reagents

Catalog No. 23666435 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
4 x 250 mL
4 x 50mL R1, 2 x 23mL R2
5 x 150 R2mL
5 x 300 R1mL
Form:
Liquid
Powder
Formulation:
4-AAP >0.2mM, HDCBS 2mM, Uricase (microbial) >150U/L, Peroxidase (horseradish) >2500U/L, pH 7.5 ± 0.1 Buffer, non-reactive Stabilizers
4-Aminoantipyrine (toxic) (dilute) 0.04%, Potassium Ferricyanide 0.025g/L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Formulation
23-666-435 4 x 250 mL Powder 4-Aminoantipyrine (toxic) (dilute) 0.04%, Potassium Ferricyanide 0.025g/L
23-666-431 5 x 300 R1mL Liquid 4-Aminoantipyrine (toxic) (dilute) 0.04%, Potassium Ferricyanide 0.025g/L
23-666-425 4 x 50mL R1, 2 x 23mL R2 Liquid 4-AAP >0.2mM, HDCBS 2mM, Uricase (microbial) >150U/L, Peroxidase (horseradish) >2500U/L, pH 7.5 ± 0.1 Buffer, non-reactive Stabilizers
23-666-432 5 x 150 R2mL Liquid 4-Aminoantipyrine (toxic) (dilute) 0.04%, Potassium Ferricyanide 0.025g/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-666-435 Supplier Pointe Scientific Supplier No. U75801L

missing translation for 'validMainframeMsg'

Includes

4-aminoantipyrine >0.3mM; TBHBA >1.0m; Uricase 150 U/L; Peroxidase >2500 U/L; Buffer; Non-reactive stabilizers and fillers

Quantify uric acid levels in serum

  • Method: Uricase/Trinder, Endpoint
  • Format: Dry powder
  • Wavelength: 520nm
  • Linearity: 25mg/dL
  • Expected Values: 2.5 to 7.7mg/dL
  • Reconstituted stability: 48 hours RT, 31 days 2 to 8°C

For the quantitative determination of Uric Acid in serum

Specifications

Certifications/Compliance CE Marked
Type Uric Acid Standard Reagent
Formulation 4-Aminoantipyrine (toxic) (dilute) 0.04%, Potassium Ferricyanide 0.025g/L
For Use With (Application) Quantitative determination of Uric Acid in Serum
Quantity 4 x 250 mL
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.