Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Uric Acid Liquid Reagents

Catalog No. 23666430 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120 mL
1 x 500 mL
1 x 1 L
Form:
Liquid
Formulation:
4-AAP >0.2mM, HDCBS 2mM, Uricase (microbial) >150U/L, Peroxidase (horseradish) >2500U/L, pH 7.5 ± 0.1 Buffer, non-reactive Stabilizers
4-Aminoantipyrine >0.3mM, TBHBA >1mM, Uricase 150U/L, Peroxidase >2500U/L, Buffer, non-reactive Stabilizers and fillers
pH:
7.5
8.1
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Formulation pH
23-666-430 1 x 500 mL Liquid 4-AAP >0.2mM, HDCBS 2mM, Uricase (microbial) >150U/L, Peroxidase (horseradish) >2500U/L, pH 7.5 ± 0.1 Buffer, non-reactive Stabilizers 7.5
23-666-428 1 x 120 mL Liquid 4-Aminoantipyrine >0.3mM, TBHBA >1mM, Uricase 150U/L, Peroxidase >2500U/L, Buffer, non-reactive Stabilizers and fillers 8.1
23-666-429 1 x 1 L Liquid 4-Aminoantipyrine >0.3mM, TBHBA >1mM, Uricase 150U/L, Peroxidase >2500U/L, Buffer, non-reactive Stabilizers and fillers 8.1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-430 Supplier Pointe Scientific Supplier No. U7581500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent requires Uric Acid Standard (23-666-424)/Chemistry Calibrator (23-666-189)

Method: Uricase/trinder, endpoint
Storage Temperature: 2–8°C
Reconstituted Stability: Ready-to-use liquid
Linearity: 20mg/dL
Measurement Wavelength: 520nm
Reaction Temperature: 37°C
Expected Values: 2.5–7.7mg/dL

Specifications

Certifications/Compliance CE Marked
Type Uric Acid Reagent Set
Formulation 4-AAP >0.2mM, HDCBS 2mM, Uricase (microbial) >150U/L, Peroxidase (horseradish) >2500U/L, pH 7.5 ± 0.1 Buffer, non-reactive Stabilizers
For Use With (Application) Quantitative determination of Uric Acid in Serum, in-vitro diagnostic use only
pH 7.5
Quantity 1 x 500 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.