Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Uric Acid, Xanthine Oxidase Product, >99% Purity, 10 g

Catalog No. 502704805
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4805 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4805 Supplier Abcam Supplier No. AB14624310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Final oxidation product of purine catabolism that acts as a potent peroxynitrite scavenger and antioxidant.

  • Molecular weight: 168.11 g/mol
  • Purity: >99%
  • Description: Xanthine oxidase product; final oxidation product of purine catabolism in humans and primates.
  • Application: Potent peroxynitrite scavenger and antioxidant
  • Storage buffer: 100% Uric acid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Antioxidant
Content And Storage Store at ambient temperature.
Buffer 100% Uric acid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Uric Acid
Formulation 100% Uric acid
Quantity 10 g
For Use With (Application) Peroxynitrite scavenging, Antioxidant activity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.