Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Uricase (C. utilis)

Catalog No. 89966727 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-727 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-966-727 Supplier Abnova™ Supplier No. P5261
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) ∽120kDa
Name Uricase (C. utilis)
Preparation Method Native protein purified from Candida utilis
Quantity 100 U
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name URICASE
Biological Activity ≥2 units per mg dry weight. One unit oxidizes one micromole of uric acid per minute at 25°C and pH 8.5 under the specified conditions.
Species Yeast
Protein Tag None
Expression System Native protein purified from Candida utilis
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.