Learn More
Medchemexpress LLC Uridine 5'-monophosphate | 58-97-9 | 99.8% | 324.18 g/mol | C9H13N2O9P | 100 MG

Supplier: Medchemexpress LLC HY101981R100MG
Uridine 5'-monophosphate (free acid) is an analytical standard used as a reference compound in biochemical assays, chromatography, and mass spectrometry. It is applied in studies of nucleotide metabolism and as a calibration standard in analytical workflows. The material is supplied as a high-purity solid suitable for research applications.
- Purity 99.8%.
- Molecular weight 324.18 g/mol.
- Molecular formula C9H13N2O9P.
- Appearance solid, white to off-white.
- Pack size 100 MG.
- Storage follow recommended conditions in the certificate of analysis.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.