Learn More
Medchemexpress LLC Uridine triphosphate trisodium dihydrate | 116295-90-0 | MFCD00150714 | 97.0% | 586.12 g/mol | C9H16N2Na3O17P3 | 100 MG

Supplier: Medchemexpress LLC HY107372A100MG
Uridine triphosphate trisodium dihydrate is the trisodium salt dihydrate form of UTP, supplied as a solid nucleotide reagent for in vitro biochemical and molecular biology applications. It functions as a substrate for enzymatic assays and nucleotide incorporation reactions and is provided with specification and storage guidance for research use.
- Purity 97.0%.
- Physical form solid dihydrate.
- Water solubility 116.67 mg/mL (199.05 mM); may require ultrasonic agitation.
- Molecular weight 586.12 g/mol.
- Storage sealed at -20°C for solid; in solution store at -80°C up to 6 months.
- Intended use in enzymatic assays and nucleotide incorporation reactions in vitro.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.