Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG™ Urinal Grippy™ Mat
SDP

Catalog No. 18366547 Shop All New Pig Corporation Products
Change view
Click to view available options
Dimensions (L x W):
23.5 x 19 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-366-547 23.5 x 19 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-547 Supplier PIG™ Supplier No. GRP7002BK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keep your restrooms cleaner and provide better drip and splash protection with our adhesive-backed urinal mats.

  • Antimicrobial technology prevents the growth of odor-causing bacteria and mold on the mats to keep restrooms cleaner and fresher smelling
  • Proprietary, full-coverage adhesive backing keeps mats in place underfoot with no shifting, curling or flipping up to stop slip and trip hazards; peels up easily
  • Mats stick to your floor and won't budge while you clean them
  • Liquidproof barrier layer stops liquid pass-through to prevent stains and grout damage
  • Tested and certified by the National Floor Safety Institute
  • For indoor use on concrete, vinyl, linoleum, tile and laminate flooring
  • 19 x 23.5 in.
  • 6 pads per box

Specifications

Dimensions (L x W) 23.5 x 19 in.
Length (English) 23.5 in.
Type Urinal Mat
Color Black
Material Plastic
Length (Metric) 60 cm
Width (English) 19 in.
Width (Metric) 48 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.