Learn More
Description
Students will:
- Use simulated urine to avoid the unpleasantness of using the real thing
- Examine simulated urine factors such as pH, color and clarity, as well as test for the presence or absence of proteins, glucose and calcium
- Examine samples microscopically to determine if any crystals are present in the sample
Specifications
Specifications
| Color | White |
| Product Type | Urinalysis Kit |
| Grade | 7 to 14 |
| Age | 13 to 20 |
| For Use With (Application) | Use simulated urine to avoid the unpleasantness of using the real thing, examine simulated urine factors such as ph, color and clarity, as well as test for the presence or absence of proteins, glucose and calcium |
| Includes | Teacher's guide, student study guide copymasters, kit with simulated urine samples (250 mL each of patient X, patient Y, patient Z, control), 25 mL Benedict's reagent, 25 mL biuret solution, 25 mL Sulkowitch's reagent, pH test strips, microscope slides, coverslips, 30 mL graduated Plastic cups, 1 mL graduated pipets |
| Quantity | 1 Ea. |
| Dimensions (L x W x H) | 12 x 9.25 x 6.5 in. |
| Class Size | 15 Lab Groups |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
