Learn More
Zymo Research Corporation Sample Urine Conditioning Buffer (8 ml)
Supplier: Zymo Research Corporation D306118S
Free Sample Features- Effectively preserves DNA and RNA in urine atambient temperatures- Facilitates pelleting of both cellular and cell-freenucleic acids from large volume urine samples- Inhibits microbial growth during long-term (cold-free)storage of urine samples
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.