Learn More
SA Scientific Urine hCG, Serum/Urine hCG, and Ultra™ hCG Serum/Urine Tests
Catalog No. SACOL087413
Change view
Click to view available options
Quantity:
3 x 5 mL
30 Tests
50 Tests
Type:
Serum Control Set
Serum, Urine
Ultra Serum Control Set
Ultra Serum/Urine Test
hCG Urine Control
hCG Urine Test
Sample Type:
Serum
Serum, Urine
Urine
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Sample Type
SACOL087413 3 x 5 mL hCG Urine Control Urine
SACOL087750 50 Tests Serum, Urine Serum, Urine
SACOL087731 30 Tests Ultra Serum/Urine Test Serum, Urine
SACOL087751 50 Tests Ultra Serum/Urine Test Serum, Urine
SACOL087712 3 x 5 mL Serum Control Set Serum
SACOL087715 3 x 5 mL Ultra Serum Control Set Serum
SACOL087430 30 Tests hCG Urine Test Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. SACOL087413 Supplier SA Scientific Supplier No. COL087413
Add to Cart
Add to Cart
Results in as little as four minutes
  • CLIA Complexity: Waived for Urine Only
  • CPT Codes: 81025 (Waived), 84703 (Non-waived)
  • Available with choice of sensitivity—10 or 20mIU/mL in Ultra Serum/Urine or 25mIU/mL in Serum/Urine and urine only tests
  • Visual, one-step test for the qualitative determination of human chorionic gonadotropin (hCG) in serum or urine to aid in early detection of pregnancy
  • Results in four to eight minutes
  • Utilizes combination of monoclonal and polyclonal antibody reagent to selectively detect elevated levels of hCG
  • Positive specimens react with the specific antibody-hCG colored conjugate and form a colored line in the S (specimen) area; absence of this colored line suggests a negative result
  • A colored line in the C (control) area will always appear regardless of the presence or absence of hCG
  • Addition of LH (300mIU/mL), FSH (1000mIU/mL) and TSH (1000μIU/mL) to negative (0mIU/mL hCG) or positive (25mIU/mL hCG) serum/urine samples showed no cross-reactivity
Specifications
CPT Code 81025
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Urine
Type hCG Urine Control
Includes Negative, low positive and high positive control
Quantity 3 x 5 mL
Test Time 4 to 8 min.