Learn More
CPC Scientific H-Phe-Thr-Leu-Ser-Leu-Asp-Val-Pro-Thr-Asn-Ile-Met-Asn-Leu-Leu-Phe-Asn-Ile-Ala-Lys-Ala-Lys-Asn-Leu-Arg-Ala-Gln-Ala-Ala-Ala-Asn-Ala-His-Leu-Met-Ala-Gln-Ile-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific UROC004A
Encompass
A highly selective ligand for the CRF-II receptor, this peptide features an amino acid sequence of the urocortin III peptide that corresponds to amino acids three to 40 of stresscopin (human).ONE-LETTER SEQUENCE: FTLSLDVPTNIMNLLFNIAKAKNLRAQAAANAHLMAQI-NH2MOLECULAR FORMULA: C185H307N53O50S2MOLECULAR WEIGHT:4137.93STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [357952-09-1], SYNONYMS: Stresscopin (3-40) (human)RESEARCH AREA: NeuropeptidesREFERENCES: K.Lewis et al., Proc. Natl. Acad. Sci. USA , 98, 7570 (2001)
Catalog No. 50-211-0592