Learn More
CPC Scientific H-Phe-Thr-Leu-Ser-Leu-Asp-Val-Pro-Thr-Asn-Ile-Met-Asn-Leu-Leu-Phe-Asn-Ile-Ala-Lys-Ala-Lys-Asn-Leu-Arg-Ala-Gln-Ala-Ala-Ala-Asn-Ala-His-Leu-Met-Ala-Gln-Ile-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific UROC004B
Encompass
A highly selective ligand for the CRF-II receptor, this peptide features an amino acid sequence of the urocortin III peptide that corresponds to amino acids three to 40 of stresscopin (human).ONE-LETTER SEQUENCE: FTLSLDVPTNIMNLLFNIAKAKNLRAQAAANAHLMAQI-NH2MOLECULAR FORMULA: C185H307N53O50S2MOLECULAR WEIGHT:4137.93STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [357952-09-1], SYNONYMS: Stresscopin (3-40) (human)RESEARCH AREA: NeuropeptidesREFERENCES: K.Lewis et al., Proc. Natl. Acad. Sci. USA , 98, 7570 (2001)
Catalog No. 50-211-0593
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.