Learn More
CPC Scientific H-Thr-Ala-Pro-Arg-Ser-Leu-Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Met-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NATR004B
Encompass
Urodilatin (95-126) originally extracted from human urine that corresponds to the family of natriuretic-vasorelaxant peptides found earlier in heart atria. Infusion of this peptide is found to represent a concept for the treatment of therapy-resistant acute renal failure after liver transplantion. The use of NATR-004 is protected by patents. ONE-LETTER SEQUENCE: TAPRSLRRSSCFGGRMDRIGAQSGLGCNSFRY (C11&C27 bridge)MOLECULAR FORMULA: C145H234N52O44S3MOLECULAR WEIGHT:3506STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [115966-23-9], RESEARCH AREA: CardiovascularREFERENCES: P. Schultz-Knappe et al., Klin. Wochenschr., 66, 752 (1988)
Catalog No. 50-210-9854