Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urokinase, human urine, Thermo Scientific™

Catalog No. AAJ60553LB0
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60553LB0 5 mg
AAJ60553MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60553LB0 Supplier Thermo Scientific Chemicals Supplier No. J60553LB0

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9039-53-6
MDL Number MFCD00132566
Synonym EC 3.4.21.73

Specifications

Packaging Plastic bottle
Quantity 5 mg
Solubility Information Reconstitute in water at 100&&Num;181;g/ml
Physical Form Lyophilized Powder
Chemical Name or Material Urokinase
Hazard Category H300
Precautionary Statement P264b-P270-P301+P310-P330-P501c
EINECSNumber 232-917-9
RTECSNumber OB8900000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.