Learn More
CPC Scientific H-Asn-Asp-Asp-Pro-Pro-Ile-Ser-Ile-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Asn-Met-Ile-Glu-Met-Ala-Arg-Ile-Glu-Asn-Glu-Arg-Glu-Gln-Ala-Gly-Leu-Asn-Arg-Lys-Tyr-Leu-Asp-Glu-Val-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific UROT001B
Encompass
Homologous sequence-containing peptide with hypothalamic corticotropin-releasing factor (CRF) and the frog skin peptide sauvagine.ONE-LETTER SEQUENCE: NDDPPISIDLTFHLLRNMIEMARIENEREQAGLNRKYLDEV-NH2MOLECULAR FORMULA: C210H340N62O67S2MOLECULAR WEIGHT:4869.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [83930-33-0], RESEARCH AREA: NeuropeptidesREFERENCES: K. Lederis et al., Science, 218, 162 (1982)
Catalog No. 50-211-0599