Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ursolic acid, 95%

Catalog No. p-4510425
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC290182500 250 mg Glass bottle
AC290180010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC290182500 Supplier Thermo Scientific Chemicals Supplier No. 290182500
Only null left
Add to Cart

CAS: 77-52-1 | C30H48O3 | 456.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-52-1
Molecular Formula C30H48O3
Molecular Weight (g/mol) 456.71
MDL Number MFCD00009621
InChI Key WCGUUGGRBIKTOS-GPOJBZKASA-N
Synonym 3beta-Hydroxyurs-12-en-28-oic acid
PubChem CID 129316832
IUPAC Name (4aS,6aS,6aS,6bR,8aS,12aR,14bR)-10-hydroxy-1,2,6a,6b,9,9,12a-heptamethyl-2,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydro-1H-picene-4a-carboxylic acid
SMILES C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(O)=O

Specifications

Melting Point 283.0°C to 285.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Beilstein 10,202
Merck Index 15,10075
Specific Rotation Condition + 59.00 (20.00°C c=0.3,pyridine)
Specific Rotation + 59.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in 2% alcoholic naoh,soluble in hot glacial acetic acid,moderately soluble in acetone,slightly soluble in alcohol,ether,chloroform and,carbon disulfide
Formula Weight 456.7
Percent Purity 95%
Physical Form Crystalline Powder or Needles
Chemical Name or Material Ursolic acid
Show More Show Less
EINECSNumber 201-034-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.