Learn More
CPC Scientific H-Ile-Pro-Leu-Arg-Gly-Ala-Phe-Ile-Asn-Gly-Arg-Trp-Asp-Ser-Gln-Cys-His-Arg-Phe-Ser-Asn-Gly-Ala-Ile-Ala-Cys-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ATMP009B
Encompass
Urumin is a host defense peptide isolated from the skin of the South Indian frog that shows antiviral activity (virucide) for H1 hemagglutinin-bearing human influenza A viruses. The peptide, that shows no homology to known peptide families, targets the H1 hemagglutinin protein of the conserved stalk region.SEQUENCE: H-Ile-Pro-Leu-Arg-Gly-Ala-Phe-Ile-Asn-Gly-Arg-Trp-Asp-Ser-Gln-Cys-His-Arg-Phe-Ser-Asn-Gly-Ala-Ile-Ala-Cys-Ala-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: IPLRGAFINGRWDSQCHRFSNGAIACAMOLECULAR FORMULA: C129H198N42O35S2MOLECULAR WEIGHT: 2961.35STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: AntimicrobialREFERENCES:Holthausen, David J., et al. "An Amphibian Host Defense Peptide Is Virucidal for Human H1 Hemagglutinin-Bearing Influenza Viruses." Immunity 46 (2017): 587-595.SKU(s): ATMP-009A, ATMP-009B
Catalog No. 50-210-8825
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.