Learn More
Innovative Research Inc US Cow Rumen Fluid Fz 1L
Supplier:  Innovative Research Inc IGBORFLZ1000ML
Encompass_Preferred
Innovative Grade US Origin Bovine Rumen Fluid Frozen 1000ml - Innovative Grade US Origin Bovine Rumen Fluid from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4569
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.