Learn More
MCR Safety Cambridge Protective Goggles
Cambridge Goggle offers the ultimate viewing area and maximum comfort in a lightweight design. It features a clear frame and a clear anti-fog polycarbonate lens for maximum viewing area and enhanced peripheral vision.
Supplier:  MCR Safety CM110AFN
Catalog No. 19-825-539
This item is not returnable. View return policy
Description
Description
Ergonomic shape and form fitting design prevents intrusion and can be worn over Rx eyewear, and with half- mask respirators. Distortion-free polycarbonate lenses block 99.9% of harmful UVA and UVB rays and meets the ANSI Z87+ eyewear standard.
  • Provides splash and impact protection, Neoprene strap
  • Translucent body for enhanced peripheral vision
  • Can be worn over Rx eyewear, and with half- mask respirators
  • Clear Anti-Fog abrasion resistant lens coating
  • Neoprene Strap contains No Latex
  • Meets all ANSI Z87+ eyewear requirements
Compliance
Meets ANSI Z87+ standards.
Specifications
Specifications
Clear
Neoprene strap
Indirect vent
Protective Goggles
1 Pair
Polycarbonate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.