Learn More
Hach Company USB and A/C Power Adapter kit, HQd Series Meters
Supplier:  Hach Company 5826300
Encompass_Preferred
For use with any HQd Meter, this adapter provides A/C power in the lab. The USB adapter that is included can be used to transfer data from the meter to a PC, memory stick, or printer. NOTE: Meter will remain water-proof after using the A/C power adapter. Included with HQ40d meter systems.
Catalog No. 50-199-7181
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.