Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ USB-C to RJ50 Cable for V2200 Wired Barcode Scanner

Catalog No. 19029969 Shop All Brady Corporation Products
Change view
Click to view available options
Length (English):
6.56 ft.
9.84 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-029-969 9.84 ft.
19-029-968 6.56 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-029-969 Supplier Brady™ Supplier No. 178575
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with the V2200 Wired Barcode Scanner.

Replacement cable for use with the V2200 Wired Barcode Scanner.

Available as 2 m straight cable or 3 m coiled cable.

Specifications

Length (English) 9.84 ft.
Length (Metric) 3 m
Type Cable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.