Learn More
Lascar Electronics Inc Temperature and Humidity Data Logger with USB and Display
SDP
Supplier:  Lascar Electronics Inc ELUSBTCLCD
Encompass_Preferred
This standalone data logger measures over 16,000 readings over a -35 to +80°C (-31 to +176°­­F) and 0 to 100% humidity (RH) range. At the touch of a button, use the on-board display to cycle between the current, minimum and maximum temperature logs seen during the session for both temperature and humidity. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery and wall mount clip.
Catalog No. NC0504837
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.