Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ USDA Cattle/Swine Screw-top Bottle USDA25BPW, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S26862 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
100/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S26862 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S26862 Supplier 3M™ Supplier No. USDA25BPW
Add to Cart
Add to Cart

Contains 25 mL buffered peptone water broth for 3M™ USDA cattle/swine kits

  • Cattle/swine diluent formulated for USDA testing
  • Meets the requirements of the US Pathogen Reduction Act

Specifications

Broth Type Buffered Peptone Water Broth
Cap Type Screw
Form Prepared Medium
Media Type Buffered Peptone Water Broth
Product Type Cattle/Swine Bottle
For Use With (Application) USDA cattle/swine testing
For Use With (Equipment) 3M™ USDA cattle/swine kits
Includes 25 mL buffered peptone water broth
Packaging Type Bottle
Product Line Neogen
Quantity 100/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.