Learn More
Bulbman Inc USHIO 6 Volt, 15 Watt, Incandescent 8018 Bulb
SDP
Supplier:  Bulbman Inc 108768
Encompass_Preferred
6 Volt 15watt Incandescent - Double Contact Bayonet Base - 100 Hour Life
Catalog No. NC9066446
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.