Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP 467 Calibration Mixture #4, Restek™
SDP

Catalog No. 06802563 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-802-563 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-802-563 Supplier Restek Corporation Supplier No. 36006
May include imposed supplier surcharges.
Add to Cart
Add to Cart
  • The Class 1 mixtures below reflect the requirements of USP23/NF18 effective January 1, 1995 to December 31, 1999.
  • Mixtures do not meet the current USP guidelines, many still use these mixtures to obtain a detectable benzene peak for the direct injection methods, Method I and Method V
Components
  • Benzene (71-43-2), 2μg/mL
  • Chloroform (67-66-3), 60μg/mL
  • 1,4-Dioxane (123-91-1), 380μg/mL
  • Methylene chloride (dichloromethane) (75-09-2), 600μg/mL
  • Trichloroethene (79-01-6), 80μg/mL

 

Specifications

Chemical Name or Material Methanol
CAS Max % 99.887800
Ignition Point 464°C
Odor Mild
Vapor Density 1.1 (Air = 1)
Flash Point 52°C
Specific Gravity 0.791 - 0.792 g/cm3 at 20°C
Molecular Formula CH4O
Concentration In methanol, 1 mL/ampul
MDL Number MFCD00004595
Melting Point -98°C
Type Multi-component Standard
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Quantity 1 mL
Formula Weight 32.04
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.