Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lime Media USP Grade Sanitizer - 70% Ethanol

Catalog No. 19043996
Change view
Click to view available options
Packaging:
Case (4)
Pallet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
19-043-996 Case (4)
19-043-997 Pallet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-043-996 Supplier Lime Media Supplier No. 197771UE
This item is not returnable. View return policy

Formula meets requirements outlined by the United States Pharmacopeia (USP)

  • USP Grade Sanitizer
  • 70% Ethanol
  • 300 Viscosity
  • 1 Gallon Bottles

Specifications

Product Type Sanitizing Fluid
Composition 70% Ethanol
Packaging Case (4)
Quantity 4/Cs.
Volume (English) 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.