Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP14 Monoclonal Antibody (889CT6.2.1)
GREENER_CHOICE

Catalog No. PIMA524727
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524727 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524727 Supplier Invitrogen™ Supplier No. MA524727
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

USP14 Monoclonal Antibody for Western Blot, ICC/IF

This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP14
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 889CT6.2.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Usp14
Gene Accession No. P54578, Q9JMA1
Gene Alias 2610005K12Rik; 2610037B11Rik; ataxia; AW107924; ax; C78769; deubiquitinating enzyme 14; dUB-type TGT for deubiquitinating enzyme; nmf375; TGT; tRNA-guanine transglycosylase, 60-kD subunit; ubiquitin carboxyl-terminal hydrolase 14; ubiquitin specific peptidase 14; ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase); ubiquitin specific protease 14; ubiquitin specific protease 14 (tRNA-guanine transglycosylase); ubiquitin thioesterase 14; ubiquitin thiolesterase 14; ubiquitin-specific processing protease 14; ubiquitin-specific-processing protease 14; Usp14; wu:fk63d09; zgc:55949
Gene Symbols Usp14
Host Species Mouse
Immunogen Purified His-tagged USP14 protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291796, 59025, 9097
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.