Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP14 Recombinant Rabbit Monoclonal Antibody (3A3J1)
GREENER_CHOICE

Catalog No. PIMA542711
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA542711 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIMA542711

Supplier: Invitrogen™ MA542711

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, HCT116, U-251MG, Mouse brain, Rat Brain. Immunogen sequence: RSKFKDLEDK KVNQQPNTSD KKSSPQKEVK YEPFSFADDI GSNNCGYYDL QAVLTHQGRS SSSGHYVSWV KRKQDEWIKF DDDKVSIVTP EDILRLSGGG DWH.

This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP14
Applications ELISA, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 3A3J1
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Usp14
Gene Accession No. P54578, Q9JMA1
Gene Alias 2610005K12Rik; 2610037B11Rik; ataxia; AW107924; ax; C78769; deubiquitinating enzyme 14; dUB-type TGT for deubiquitinating enzyme; nmf375; TGT; tRNA-guanine transglycosylase, 60-kD subunit; ubiquitin carboxyl-terminal hydrolase 14; ubiquitin specific peptidase 14; ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase); ubiquitin specific protease 14; ubiquitin specific protease 14 (tRNA-guanine transglycosylase); ubiquitin thioesterase 14; ubiquitin thiolesterase 14; ubiquitin-specific processing protease 14; ubiquitin-specific-processing protease 14; Usp14; wu:fk63d09; zgc:55949
Gene Symbols Usp14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 371-473 of human USP14 (P54578).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291796, 59025, 9097
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.