Learn More
TARGETMOL CHEMICALS INC USP30 INHIBITOR 18 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T366825MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. USP30 inhibitor 18 is a selective inhibitor of USP30 (IC50 = 0.02 uM). USP30 inhibitor 18 is able to accelerate mitophagy and increase protein ubiquitination. purity: 99%
Catalog No. 50-258-0171