Learn More
Invitrogen™ USP37 Polyclonal Antibody

Description
Antibody detects endogenous levels of total USP37.
Specifications
Specifications
| Antigen | USP37 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Usp37 |
| Gene Accession No. | Q86T82 |
| Gene Alias | 4932415L06Rik; C330008N13Rik; Deubiquitinating enzyme 37; Kiaa1594; tmp_locus_50; ubiquitin carboxyl-terminal hydrolase 37; ubiquitin specific peptidase 37; ubiquitin specific peptidase 37-like; ubiquitin specific protease 37; ubiquitin thioesterase 37; ubiquitin thiolesterase 37; ubiquitin-specific-processing protease 37; USP37 |
| Gene Symbols | Usp37 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.