Learn More
Invitrogen™ USP40 Monoclonal Antibody (OTI2C5)

Description
USP40 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | USP40 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2C5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | USP40 |
| Gene Accession No. | Q9NVE5 |
| Gene Alias | B230215L03Rik; C730029K03; deubiquitinating enzyme 40; LRRGT00071; Ubiquitin carboxyl-terminal hydrolase 40; ubiquitin specific peptidase 40; ubiquitin specific protease 40; ubiquitin thioesterase 40; ubiquitin thiolesterase 40; ubiquitin-specific-processing protease 40; USP40 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.