Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566720
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566720 Supplier Invitrogen™ Supplier No. PA566720
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TIVDKASESSD PSAYQNQPGS SEAVSPGDMD AGSASWGAVS SLNDVSNHTL SLGPVPGAVV YSSSSVPDKS KPSPQKDQAL GDGIAPPQKV L Highest antigen sequence identity to the following orthologs - mouse 83%, rat 85%.

USP42 (ubiquitin specific peptidase 42) is a 1,325 amino acid protein that belongs to the peptidase C19 family. Expressed in a variety of tissues, USP42 functions to catalyze the conversion of a ubiquitin C-terminal thioester to a free ubiquitin and a thiol. The gene encoding USP42 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP42
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Usp42
Gene Accession No. Q9H9J4
Gene Alias 2410140K03Rik; 3110031A07Rik; A630018G05Rik; D5Ertd591e; Deubiquitinating enzyme 42; ubiquitin carboxyl-terminal hydrolase 42; ubiquitin specific peptidase 42; ubiquitin specific protease 42; ubiquitin thioesterase 42; ubiquitin thiolesterase 42; ubiquitin-specific-processing protease 42; Usp42
Gene Symbols Usp42
Host Species Rabbit
Immunogen Recombinant Human USP42. Recombinant protein control fragment (Product #RP-105344).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84132
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.