Learn More
USP47 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301048AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Putative ubiquitin-specific-processing protease that regulates cell growth and survival. Probably regulates CDC25A expression at a transcriptional level. May be catalytically inactive although it seems to have kept all necessary active site residues.Specifications
| USP47 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q96K76 | |
| USP47 | |
| Between 1 and 50 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| USP47 | |
| TRFP, Trf (TATA binding protein-related factor)-proximal homolog, deubiquitinating enzyme 47, ubiquitin carboxyl-terminal hydrolase 47, ubiquitin specific protease 47, ubiquitin thioesterase 47, ubiquitin thiolesterase 47, ubiquitin-specific-processing protease 47 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55031 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.