Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP6NL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558691
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558691 Supplier Invitrogen™ Supplier No. PA558691
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRQYNHAAAN QNSNATSNIR KEFVPKWNKP SDVSATERTA KYTMEGKGRA AHPALAVTVP GPAEVRVSNV RPKMKALDAE DGKRGSTAS Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 66%.

Acts as a GTPase-activating protein for RAB5A and RAB43. Involved in receptor trafficking. In complex with EPS8 inhibits internalization of EGFR. Involved in retrograde transport from the endocytic pathway to the Golgi apparatus. Involved in the transport of Shiga toxin from early and recycling endosomes to the trans-Golgi network. Required for structural integrity of the Golgi complex. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP6NL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USP6NL
Gene Accession No. Q92738
Gene Alias AI316785; Kiaa0019; mKIAA0019; novel TBC domain containing protein; related to the N-terminus of tre; RNTRE; RN-tre; TRE2NL; USP6 N-terminal like; USP6 N-terminal-like protein; Usp6nl; USP6NL-IT1
Gene Symbols USP6NL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USP6NL. Recombinant protein control fragment (Product #RP-97098).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9712
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.