Learn More
World Precision Instrument Ussing System for Epithelial Research-USS7S
SDP
Supplier:  World Precision Instrument USS7S
Encompass
WPI’s classical Ussing Chambers are well established perfusion chambers that are easy to operate, easy to control temperature, and easy to clean after use. Ussing Cham­bers are machined from solid acrylic with eight entry ports for fluid lines, elec­trodes, or agar bridges. For easy, leak-free attachment of tubing and elec­trodes, all eight ports are Luer type. The four ports for volt­age and current electrodes are re­cessed to prevent formation of air bubbles in the chamber. The fluid com­part­ments in each side of the chamber are sep­a­rat­ed by the epithelial membrane being stud­ied. Sharp stainless steel pins on one side of the chamber hold the mem­brane in po­si­tion and mate with holes in the opposite chamber in­ter­face. (In the CHM4, tissue is held by an O-ring instead of pins.)
Catalog No. 50-252-1034
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.