Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTF1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7188917
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3270 10 μL
50-199-3272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3270 Supplier Active Motif Supplier No. 61254
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Undifferentiated transcription factor-1 (UTF-1) is used as marker for the undifferentiated state of pluripotent stem cells.UTF1 is a chromatin-associated protein with core histone-like properties. UTF1 further acts as a transcriptional repressor and is required for proper differentiation of pluripotent cells.

Specifications

Antigen UTF1
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene UTF1
Gene Accession No. Q5T230, Q6J1H4
Gene Alias AI505934; OTTHUMP00000020786; Undifferentiated embryonic cell transcription factor 1; Utf1
Gene Symbols UTF1
Host Species Rabbit
Immunogen This UTF1 antibody was raised against a peptide within the N-terminal region of human UTF1.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22286, 8433
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.