Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ uTIP™ CleanPak™ Reload, Sterile Low Retention Universal Pipette Tips, Standard

Catalog No. 12111016 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
300 μL
1250 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-111-016 10 μL
12-111-018 200 μL
12-111-019 300 μL
12111020 1250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 12-111-016 Supplier BIOTIX™ Supplier No. 63300062
This item is not returnable. View return policy

Designed with hermetically sealed bag to avoid contamination. Biotix™ uTIP™ CleanPak™ Reload, Low Retention, Pre-Sterilized Universal Pipette Tips reduce time to rack products, cost compared to pre-racked products, and environmental footprint.

  • Fits all standard pipettes
  • Improves ergonomics by lowering insertion and ejection forces with FlexFit and Blade technologies
  • CleanPak reload features advanced technology of Biotix tips
  • Allows re-use of previous racks
  • 960 tips on lab bench using the same surface area as 1 rack
  • Sterile and non-Sterile re-sealable bag available
  • Clear bag and color coded snap cards for quick identification
  • Fully recyclable and contains no contaminating cardboard or paper
  • Inserts are easy to transfer to Biotix empty racks
  • Sterile products undergo E-Beam radiation
  • All racks include label with unique bar-coded serial number, part number, and description for traceability to individual rack

Certifications

ISO 9001 , FDA Class I

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Refill
Tip Style Fine Point
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line uTIP CleanPak
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.