Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ uTIP™ CleanPak™ Reload, Sterile Low Retention Universal Pipette Tips, Standard

Catalog No. 12111129 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
250 μL
1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-111-129 250 μL
12-111-135 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-111-129 Supplier BIOTIX™ Supplier No. 63300065

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Easy to transfer and come in a resealable bag, free of contaminating cardboard and paper. Validated for fit and accuracy on universal pipettes.

  • Validated for fit and accuracy on standard pipettes like Eppendorf™, Gilson™ and Biohit™
  • Reduces plastic waste and bench space
  • Resaealable bag provides better protection from airborne contaminants
  • Clear bag and color coded snap cards for quick identification
  • Fully recyclable and contains no contaminating cardboard or paper
  • FlexFit™ forms a secure seal with less force, reducing risk of repetitive stress injuries
  • X-Resin™ and Blade™ provide you maximum sample recovery and precision
  • Certifications

    ISO 9001

    TRUSTED_SUSTAINABILITY

    Specifications

    Description 250 μL, Reload, Sterile
    Volume (Metric) 250 μL
    Filtered Non-filtered
    Sterility Sterile
    Color Clear
    Rack Color Green
    Autoclavable Autoclavable
    Model CleanPak Universal
    Length (Metric) 59.54 mm
    Certifications/Compliance ISO 9001
    Clean Claims Fully Recyclable
    For Use With (Application) Low binding for Protein, DNA, and nanoparticles
    Sterilization Method e-beam
    Format Refill
    Tip Style Ergonomic
    Packaging Reload
    Quantity 10 refills/pk
    Product Type Pipette Tips
    Disposable Single-use
    For Use With (Equipment) Universal Fit
    Graduated Yes
    Product Line Biotix uTip
    Material Polypropylene
    Shape Conical
    Graduations 10 μL, 50 μL, 100 μL, 200 μL
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.