Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ uTIP™ Filter Sterile Low Retention Universal Pipette Tips, Standard
GSA_VAGREENER_CHOICE

Catalog No. 12111000 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
100 μL
200 μL
300 μL
1000 μL
1250 μL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-111-000 10 μL
12-111-002 20 μL
12-111-003 100 μL
12-111-004 300 μL
12-111-005 1250 μL
12-111-126 200 μL
12-111-132 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 12-111-000 Supplier BIOTIX™ Supplier No. 63300154
Add to Cart
Add to Cart
This item is not returnable. View return policy

Experience a better universal tip. Biotix™ uTIP™ Filter Sterile Pipette Tips made with Flexfit™ creates a secure seal on various pipettes with less insertion and ejection force.

Experience a better universal tip with Biotix™ uTIP™

  • Biotix filter sterile tips offers aerosol protection to prevent cross contamination
  • Fits all standard universal shaft single channel, multichannel and electronic pipettes
  • FlexFit technology features flexible proximal end to ensure secure seal with 66 to 80% less insertion force than competitor tips
  • X-Resin drastically reduces sample retention resulting in tighter CV values
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
  • Each lot of Biotix tips are tested and certified free of RNase, DNase, Endotoxins/Pyrogens, PCR Inhibitors, trace metals, and human DNA

Certifications

ISO 9001

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Color Red
Autoclavable Autoclavable
Format Rack
Tip Style Fine Point
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Biotix uTip
Material Polypropylene
Shape Conical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.