Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTP18/WDR50 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560679
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0679 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0679 Supplier Bethyl Laboratories, Inc Supplier No. A301552AT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

UTP18 is involved in nucleolar processing of pre-18S ribosomal RNA.

Specifications

Antigen UTP18/WDR50
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene UTP18
Gene Accession No. Q9Y5J1
Gene Alias CGI-48, UTP18 small subunit (SSU) processome component homolog, UTP18 small subunit (SSU) processome component homolog (yeast), UTP18 small subunit processome component, UTP18, small subunit (SSU) processome component, homolog, WD repeat domain 50, WD repeat-containing protein 50, WDR50
Gene Symbols UTP18
Host Species Rabbit
Immunogen Between 325 and 375
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51096
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.