Learn More
Vector Laboratories UV Cleavable Biotin-Azide, 10mg
SDP
Supplier:  Vector Laboratories EVU102
Encompass
Chemical Formula: C38H51N7O9S, Molecular Weight: 781.92, Solubility: DMSO, MeOH, The UV Cleavable Biotin-Azide allows the tagging of alkyne modified substrates via copper-catalyzed click chemistry.
Catalog No. NC1573054
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.