Learn More
Promega UV Bulb, Maxwell 16, 1 each
Catalog No. PRSP1080 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRSP1080 1 Ea.
1 options
Catalog No. PRSP1080 Supplier Promega Supplier No. SP1080
Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
  • Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
  • Standard Elution Volume (SEV) is 200−400μl
  • Low Elution Volume (LEV) is 30−100μl
  • Plungers and Elution Tubes for the Maxwell(R) 16 Instruments only (AS2000 and AS3000)
Specifications
Quantity 1 Ea.