Learn More
Labconco UV Lamp Kit
SDP
Catalog No. 13118379 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
8 ft. Logic Biosafety Cabinet
Catalog No. For Use With (Equipment)
13-118-379 8 ft. Logic Biosafety Cabinet
1 options
Catalog No. 13-118-379 Supplier Labconco Supplier No. 3289101
Add to Cart
Add to Cart
UV lamp kit for 8 ft. Logic Biosafety Cabinets.
  • UV lamp kit for 8 ft. Logic Biosafety cabinets.
  • Includes two UV bulbs, ballast, and necessary mounting hardware.
  • Field installation required.
Specifications
Product Type UV Light
For Use With (Equipment) 8 ft. Logic Biosafety Cabinet