Learn More
Cytiva Cytiva UV Module, LED Technology, 280 nm Detection, 1 Unit
Catalog No. NC2894845 Shop All Cytiva Products
Encompass
The Cytiva UV Module is designed for seamless integration with ÄKTA start™ systems, providing essential UV detection at 280 nm. It features LED technology, eliminating warm-up time and preventing local heating, making it ideal for heat-sensitive samples. With a linear absorbance range of up to 1.5 AU and compatibility for a 2 mm flow cell, it enhances protein purification processes, setting it apart from traditional modules by ensuring immediate readiness and reliability.
Catalog No. NC2894845 Supplier Cytiva Supplier No. 29024018
The Cytiva UV Module is designed for seamless integration with ÄKTA start™ systems, providing essential UV detection at 280 nm. It features LED technology, eliminating warm-up time and preventing local heating, making it ideal for heat-sensitive samples. With a linear absorbance range of up to 1.5 AU and compatibility for a 2 mm flow cell, it enhances protein purification processes, setting it apart from traditional modules by ensuring immediate readiness and reliability.