Learn More
Syngene™ UV Safety Shield
SDP
Catalog No. 01257295 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Size:
Large
Catalog No. Size
01-257-295 Large
1 options
Catalog No. 01-257-295 Supplier Syngene™ Supplier No. UVSHIELDLARGE
Safety shield designed to offer UV protection.
  • The wearing of safety equipment according to laboratory regulations is recommended
  • Large Shield Dimensions: 14 W x 24 in. H
Specifications
Product Line Syngene
For Use With (Application) UV Protection
Product Type Safety Shield
For Use With (Equipment) GBox Chemi XX6 and XX9
Includes UV Safety Shield, 24 in. high
Size Large
Voltage 110/230 V
WARNING: Cancer - www.P65Warnings.ca.gov